missing translation for 'onlineSavingsMsg'
Learn More

Lycopene, MedChemExpress

Product Code. 16454807
Encompass_Preferred
Click to view available options
Quantity:
5 mg
10 mg
25 mg
50 mg
Unit Size:
10mg
25mg
50mg
5mg
This item is not returnable. View return policy

Product Code. 16454807

Brand: Medchem Express HYN0287

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress Lycopene is naturally occurring carotenoids found in tomato, tomato products, and in other red fruits and vegetables; exhibits antioxidant effects.

Lycopene is naturally occurring carotenoids found in tomato, tomato products, and in other red fruits and vegetables

Specifications

Chemical Name or Material Lycopene
CAS 502-65-8
Color Reddish Brown
Purity Grade Notes Research
Molecular Formula C40H56
Quantity 50 mg
Solubility Information THF : 5 mg/mL (9.31 mM; Need ultrasonic) ∣Ethanol : < 1 mg/mL (ultrasonic) (insoluble)
SMILES C/C(C)=C\CC/C(C)=C/C=C/C(C)=C/C=C/C(C)=C/C=C/C=C(/C=C/C=C(C)/C=C/C=C(CC/C=C(C)\C)\C)C
Molecular Weight (g/mol) 536.87
Formula Weight 536.87
Percent Purity 98.0%
Grade Research
Physical Form Solid
For Use With (Application) COVID-19-immunoregulation
Show More Show Less

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.