missing translation for 'onlineSavingsMsg'
Learn More
Learn More
LYAR Rabbit anti-Human, Polyclonal, Novus Biologicals™
Rabbit Polyclonal Antibody
£537.00
Specifications
| Antigen | LYAR |
|---|---|
| Dilution | Western Blot 0.04-0.4 ug/ml |
| Applications | Western Blot |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
Description
LYAR Polyclonal antibody specifically detects LYAR in Human samples. It is validated for Western BlotSpecifications
| LYAR | |
| Western Blot | |
| Unconjugated | |
| Rabbit | |
| Cytokine Research | |
| PBS, pH 7.2, 40% glycerol | |
| 55646 | |
| IgG | |
| Affinity purified |
| Western Blot 0.04-0.4 ug/ml | |
| Polyclonal | |
| Purified | |
| RUO | |
| Human | |
| cell growth-regulating nucleolar protein, FLJ20425, Ly1 reactive homolog (mouse), ZLYAR | |
| This antibody was developed against Recombinant Protein corresponding to amino acids: ACGESVKKIQVEKHVSVCRNCECLSCIDCGKDFWGDDYKNHVKCISEDQKYGGKGYEGKTHKGDIKQQAWIQKISELIKRPNVSPKV | |
| Primary | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
Vous avez repéré une opportunité d'amélioration ?Partager une correction de contenu
Correction du contenu d'un produit
Veuillez fournir vos retours sur le contenu du produit en remplissant le formulaire ci-dessous.
Nom du produit