missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ LYAR Polyclonal Antibody
Sheep Polyclonal Antibody
Brand: Invitrogen™ PA5149244
This item is not returnable.
View return policy
Description
Reconstitute at 0.2 mg/mL in sterile PBS.
Plays a role in the maintenance of the appropriate processing of 47S/45S pre-rRNA to 32S/30S pre-rRNAs and their subsequent processing to produce 18S and 28S rRNAs (PubMed:24495227). Also acts at the level of transcription regulation. Along with PRMT5, binds the gamma-globin (HBG1/HBG2) promoter and represses its expression (PubMed:25092918). In neuroblastoma cells, may also repress the expression of oxidative stress genes, including CHAC1, HMOX1, SLC7A11, ULBP1 and SNORD41 that encodes a small nucleolar RNA (PubMed:28686580). Preferentially binds to a DNA motif containing 5'-GGTTAT-3' (PubMed:25092918). Negatively regulates the antiviral innate immune response by targeting IRF3 and impairing its DNA-binding activity (PubMed:31413131). In addition, inhibits NF-kappa-B-mediated expression of proinflammatory cytokines (PubMed:31413131). Stimulates phagocytosis of photoreceptor outer segments by retinal pigment epithelial cells. Prevents nucleolin/NCL self-cleavage, maintaining a normal steady-state level of NCL protein in undifferentiated embryonic stem cells (ESCs), which in turn is essential for ESC self-renewal. [UniProt].
Specifications
| LYAR | |
| Polyclonal | |
| Unconjugated | |
| LYAR | |
| apoptosis-related protein; cell growth-regulating nucleolar protein; hypothetical protein FLJ20425; Ly1 antibody reactive; Ly1 antibody reactive clone; Ly1 antibody reactive homolog; Ly1 antibody-reactive protein; Lyar; LYAR protein; MLZ-264; PNAS-5; protein expressed in male leptotene and zygotene spermatocytes 264; ZC2HC2; ZLYAR | |
| Sheep | |
| Antigen affinity chromatography | |
| RUO | |
| 55646 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Lyophilized |
| Western Blot, Immunocytochemistry | |
| 0.2 mg/mL | |
| PBS with trehalose and no preservative | |
| Q9NX58 | |
| LYAR | |
| E. coli-derived recombinant human LYAR, Lys288-Lys379. | |
| 100 μg | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction