missing translation for 'onlineSavingsMsg'
Learn More
Learn More
LYAR Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
£381.00
Specifications
| Antigen | LYAR |
|---|---|
| Applications | Western Blot |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Host Species | Rabbit |
Description
LYAR Polyclonal specifically detects LYAR in Human samples. It is validated for Western Blot.Specifications
| LYAR | |
| Polyclonal | |
| Rabbit | |
| Cytokine Research | |
| cell growth-regulating nucleolar protein, FLJ20425, Ly1 reactive homolog (mouse), ZLYAR | |
| LYAR | |
| IgG |
| Western Blot | |
| Unconjugated | |
| RUO | |
| Q9NX58 | |
| 55646 | |
| Synthetic peptides corresponding to LYAR(Ly1 antibody reactive homolog (mouse)) The peptide sequence was selected from the middle region of LYAR. Peptide sequence EQQGEVKKNKRERKEERQKKRKREKKELKLENHQENSRNQKPKKRKKGQE. | |
| Primary |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title