missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ GAA Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA529051
This item is not returnable.
View return policy
Description
Recommended positive controls: 293T, A431, HeLa, HepG2. Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
This gene encodes acid alpha-glucosidase, which is essential for the degradation of glycogen to glucose in lysosomes. Different forms of acid alpha-glucosidase are obtained by proteolytic processing. Defects in this gene are the cause of glycogen storage disease II, also known as Pompe's disease, which is an autosomal recessive disorder with a broad clinical spectrum. Three transcript variants encoding the same protein have been found for this gene.
Specifications
| GAA | |
| Polyclonal | |
| Unconjugated | |
| Gaa | |
| 70 kDa lysosomal alpha-glucosidase; 76 kDa lysosomal alpha-glucosidase; acid (Pompe disease, glycogen storage disease type II); acid alpha-glucosidase; acid maltase; Aglucosidase alfa; E430018M07Rik; Gaa; glucosidase alpha, acid; glucosidase, alpha, acid; glucosidase, alpha; acid (Pompe disease, glycogen storage disease type II); LYAG; Lysosomal alpha-glucosidase | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 2548 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot | |
| 0.76 mg/mL | |
| PBS with 20% glycerol and 0.025% ProClin 300; pH 7 | |
| P10253 | |
| Gaa | |
| Recombinant protein encompassing a sequence within the C-terminus region of human LYAG. The exact sequence is proprietary. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction