missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CD229 Mouse anti-Human, APC, Clone: HLy9.25, Invitrogen™
Mouse Monoclonal Antibody
Brand: Invitrogen A15722
This item is not returnable.
View return policy
Description
CD229 (Ly9) is a cell surface receptor of the CD150 family, which includes also e.g. CD48 and CD224. Receptors of this family regulate cytokine production and cytotoxicity of lymphocytes and NK cells. High levels of CD229 are found on T and B cells, where its expression increases during their maturation. It is absent on granulocytes, bone marrow-derived dendritic cells, platelets and erythrocytes. CD229 has been also reported on mouse monocytes and NK cells. CD229 interacts homophilically through its N-terminal domain and localizes to the contact site between T cells and antigen presenting B cells during antigen-dependent immune synapse formation.
Specifications
| CD229, Ly9 | |
| Monoclonal | |
| APC | |
| LY9 | |
| RP11-312J18.1, CD229, SLAMF3, hly9, mLY9, T-lymphocyte surface antigen Ly-9, cell surface molecule Ly-9, cell-surface molecule Ly-9 | |
| Mouse | |
| Purified | |
| RUO | |
| 4063 | |
| 4° C | |
| Liquid |
| Flow Cytometry | |
| HLy9.25 | |
| PBS with 0.2% BSA and 15mM sodium azide; pH 7.4 | |
| Q9HBG7 | |
| LY9 | |
| Human CD229 | |
| 100 Tests | |
| Primary | |
| Human | |
| Antibody | |
| IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction