Learn More
Ly6K Antibody (CL2433) [Alexa Fluor™ 532], Novus Biologicals™
Mouse Monoclonal Antibody
Brand:  Novus Biologicals NBP2-36763AF532
Product Code. 30305402
  • £558.00 / 0.10mL
Description
Ly6K Monoclonal antibody specifically detects Ly6K in Human samples. It is validated for Immunohistochemistry,Western Blot,Immunohistochemistry (Paraffin)
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Ly6K
Monoclonal
Alexa Fluor 532
cancer/testis antigen 97, CO16, CT97, FLJ35226, HSJ001348, Ly-6K, lymphocyte antigen 6 complex, locus K, lymphocyte antigen 6K
This antibody was developed using a recombinant protein derived from Q17RY6, with the exact immunogen sequence remaining proprietary.
0.1 mL
Primary
Human
Purified
Immunohistochemistry, Western Blot, Immunohistochemistry (Paraffin)
CL2433
50mM Sodium Borate
Mouse
Protein A purified
RUO
54742
Store at 4⪚C in the dark.
IgG1