missing translation for 'onlineSavingsMsg'
Learn More

Ly6K Antibody, Alexa Fluor™ 700, Novus Biologicals™

Product Code. 18404298 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18404298 0.1 mL 0.1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18404298 Supplier Novus Biologicals Supplier No. NBP224405AF700

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

Ly6K Polyclonal antibody specifically detects Ly6K in Human samples. It is validated for Western Blot, ELISA
TRUSTED_SUSTAINABILITY

Specifications

Antigen Ly6K
Applications Western Blot, ELISA, Immunohistochemistry (Paraffin)
Classification Polyclonal
Conjugate Alexa Fluor 700
Dilution Western Blot, ELISA
Formulation 50mM Sodium Borate
Gene Alias cancer/testis antigen 97, CO16, CT97, FLJ35226, HSJ001348, Ly-6K, lymphocyte antigen 6 complex, locus K, lymphocyte antigen 6K
Host Species Rabbit
Immunogen Reacts with residues 82-100 RQRDPEDSQRTDEGDNRVW of the 14 kDa human Ly-6K protein.
Purification Method Immunogen affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Cell Cycle and Replication
Primary or Secondary Primary
Gene ID (Entrez) 54742
Target Species Human
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.