missing translation for 'onlineSavingsMsg'
Learn More
Learn More
LY6H Rabbit anti-Human, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 23933-1-AP-20UL
This item is not returnable.
View return policy
Description
LY6H is highly expressed in particular subdivisions of human brain and also in MOLT-3 and -4 acute lymphoblastic leukemia cells. It has some isoforms with MW 14-16 kDa.Specifications
| LY6H | |
| Polyclonal | |
| Unconjugated | |
| LY6H | |
| Ly 6H, LY6H, Lymphocyte antigen 6H | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 4062 | |
| -20°C | |
| Liquid |
| Western Blot, Immunohistochemistry (Paraffin) | |
| 0.3 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| O94772 | |
| LY6H | |
| LY6H Fusion Protein Ag20762 | |
| 20 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction