missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ LY6G6D Recombinant Mouse Monoclonal Antibody (9H7)

Mouse Recombinant Monoclonal Antibody
Brand: Invitrogen™ MA560793
This item is not returnable.
View return policy
Description
LY6G6D Recombinant Monoclonal Antibody for Flow, ELISA
G6D, also known as Lymphocyte antigen 6 complex locus G6D (LY6G6D), is a type I transmembrane protein belonging to the immunoglobin (Ig) superfamily, that is comprised of cell-surface proteins involved in the immune system and cellular recognition. It is a disulfide linked homodimer with an Ig-like V-type domain and is known to interact with GRB2 and GRB7 in a phosphorylation-dependent manner thus playing an important role in their downstream signal transduction pathways.
Specifications
| LY6G6D | |
| Recombinant Monoclonal | |
| 1 mg/mL | |
| PBS with 50% glycerol and 0.03% ProClin 300; pH 7.4 | |
| O95868, Q6MG58, Q9Z1Q3 | |
| LOC102119090, LY6G6D | |
| Recombinant Human LY6G6D protein. | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat, Monkey | |
| Antibody | |
| IgG1 |
| ELISA, Flow Cytometry | |
| 9H7 | |
| Unconjugated | |
| LY6G6D | |
| C6orf23; DADB-110M10.6; G6D; LY6-D; LY6G6D; Lymphocyte antigen 6 complex locus protein G6d; lymphocyte antigen 6 complex, locus G6D; lymphocyte antigen 6 family member G6D; lymphocyte antigen-6 G6D; megakaryocyte-enhanced gene transcript 1 protein; MEGT1; NG25; Protein Ly6-D | |
| Mouse | |
| Affinity chromatography | |
| RUO | |
| 102119090, 114654, 415062, 58530 | |
| -20°C or -80°C if preferred | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction