missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ LY6G6C Polyclonal Antibody

Rabbit Polyclonal Antibody

Brand:  Invitrogen™ PA524040

Product Code. 13288138

  • £479.00 / 400µL

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

LY6G6C Polyclonal Antibody for Western Blot, IHC (P)

LY6G6C belongs to a cluster of leukocyte antigen-6 (LY6)genes located in the major histocompatibility complex (MHC) classIII region on chromosome 6. Members of the LY6 superfamilytypically contain 70 to 80 amino acids, including 8 to 10cysteines. Most LY6 proteins are attached to the cell surface by aglycosylphosphatidylinositol (GPI) anchor that is directly involvedin signal transduction.
TRUSTED_SUSTAINABILITY
Specifications

Specifications

LY6G6C
Polyclonal
Unconjugated
LY6G6C
C6orf24; G6C; LY6G6C; Lymphocyte antigen 6 complex locus protein G6c; lymphocyte antigen 6 complex, locus G6C; lymphocyte antigen-6 G6C; NG24; UNQ1947/PRO4430
Rabbit
Antigen affinity chromatography, Protein A
RUO
80740
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Liquid
Immunohistochemistry (Paraffin), Western Blot
0.5 mg/mL
PBS with 0.09% sodium azide; pH 7.4
O95867
LY6G6C
KLH conjugated synthetic peptide between 62-90 amino acids from the C-terminal region of human LY6G6C.
400 μL
Primary
Human
Antibody
IgG
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.