missing translation for 'onlineSavingsMsg'
Learn More
Learn More
LY6G5C Rabbit anti-Human, Mouse, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 24633-1-AP-150UL
This item is not returnable.
View return policy
Description
11714-1-AP can detect the protein and its dimer and tetramer.
LY6G5C belongs to a cluster of leukocyte antigen-6 (LY6) genes located in the major histocompatibility complex (MHC) class III region on chromosome 6. Members of the LY6 superfamily typically contain 70 to 80 amino acids, including 8 to 10 cysteines. Most LY6 proteins are attached to the cell surface by a glycosylphosphatidylinositol (GPI) anchor that is directly involved in signal transduction.Specifications
| LY6G5C | |
| Polyclonal | |
| Unconjugated | |
| LY6G5C | |
| C6orf20, G5c, LY6G5C, LY6G5CA, LY6G5CB, NG33 | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 114652, 80741 | |
| -20°C | |
| Liquid |
| Immunoprecipitation, Western Blot | |
| 0.23 mg/mL | |
| PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
| Q5SRR4, Q8K1T5 | |
| LY6G5C | |
| LY6G5C Fusion Protein Ag18812 | |
| 150 μL | |
| Primary | |
| Human, Mouse | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction