missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Ly6g, Rat anti-Mouse, Clone: Nimp-R14, Abnova™
Rat Monoclonal Antibody
Brand: Abnova MAB16109.100ug
This item is not returnable.
View return policy
Specifications
| Ly6g | |
| Monoclonal | |
| Unconjugated | |
| In PBS (10% glycerol, 0.02% sodium azide). | |
| Gr-1/Gr1/Ly-6G | |
| Rat | |
| 100 μg | |
| Primary | |
| Mouse | |
| Antibody | |
| IgG2b |
| Blocking Assay, Flow Cytometry, Immunocytochemistry, Immunohistochemistry (Frozen), Immunohistochemistry (Paraffin) | |
| Nimp-R14 | |
| Flow Cytometry (1:100) Immunohistochemistry (Formalin/PFA-fixed paraffin-embedded sections) (1:200) Immunohistochemistry (Frozen sections) (1:200) Immunocytochemistry (1:200) Blocking (250 ug/mouse) The optimal working dilution should be determined by the end user. | |
| lymphocyte antigen 6 complex, locus G | |
| Ly6g | |
| Purified mouse BALB/c neutrophils of surface protein Ly6g. | |
| RUO | |
| 546644 | |
| For long term storage store at -20°C. Aliquot to avoid repeated freezing and thawing. | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction