missing translation for 'onlineSavingsMsg'
Learn More

LY334370, MedChemExpress

Product Code. 16463798
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 mg
10 mg
25 mg
50 mg
100 mg
Unit Size:
100mg
10mg
25mg
50mg
5mg
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16463798 10 mg 10mg
16460177 50 mg 50mg
16452866 25 mg 25mg
16416527 5 mg 5mg
16414856 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
This item is not returnable. View return policy
Product Code. 16463798 Supplier Medchem Express Supplier No. HY103107
 Restricted Item
Estimated Shipment: 11-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

MedChemExpress LY334370 is a selective 5-HT1F receptor agonist with a Ki of 1.6 nM.

LY334370 is a selective 5-HT1F receptor agonist with a Ki of 1.6 nM.

Specifications

Chemical Name or Material LY334370
CAS 182563-08-2
Color White
Purity Grade Notes Research
Molecular Formula C21H22FN3O
Quantity 10 mg
Solubility Information DMSO : ≥ 55.5 mg/mL (157.93 mM)
SMILES O=C(C1=CC=C(F)C=C1)NC2=CC3=C(C=C2)NC=C3C4CCN(C)CC4
Molecular Weight (g/mol) 351.42
Formula Weight 351.42
Percent Purity 99.8%
Grade Research
Physical Form Solid
For Use With (Application) Neuroscience-Neuromodulation
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.