missing translation for 'onlineSavingsMsg'
Learn More

LY3200882, MedChemExpress

Product Code. p-200098465
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 mg
10 mg
50 mg
100 mg
Unit Size:
100mg
10mg
50mg
5mg
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16480157 50 mg 50mg
16473778 10 mg 10mg
16416507 5 mg 5mg
16454836 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Product Code. 16480157 Supplier Medchem Express Supplier No. HY103021

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress LY3200882 is a potent, highly selective, ATP-competitive and orally active TGF-β receptor type 1 (ALK5) inhibitor with an IC50 of 38.2 nM. LY3200882 inhibits various pro-tumorigenic activities and is also used as an immune modulatory agent.

LY3200882 is a potent, highly selective, ATP-competitive and orally active TGF-β receptor type 1 (ALK5) inhibitor with an IC50 of 38.2 nM. LY3200882 inhibits various pro-tumorigenic activities and is also used as an immune modulatory agent.

Specifications

Chemical Name or Material LY3200882
CAS 1898283-02-7
Color Light Yellow
Purity Grade Notes Research
Molecular Formula C24H29N5O3
Quantity 50 mg
Solubility Information DMSO : 33.33 mg/mL (76.53 mM; Need ultrasonic)
SMILES OC(C)(C)C1=NC=CC(NC2=NC=CC(OC3=CN(C4CC4)N=C3C5CCOCC5)=C2)=C1
Molecular Weight (g/mol) 435.52
Formula Weight 435.52
Percent Purity 98.82%
Grade Research
Physical Form Solid
For Use With (Application) Cancer-Kinase/protease
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.