missing translation for 'onlineSavingsMsg'
Learn More
Learn More
LY3009120, MedChemExpress
Description
LY3009120 (DP-4978) is a pan RAF inhibitor which inhibits BRAFV600E, BRAFWT and CRAFWT with IC50s of 5.8, 9.1 and 15 nM, respectively.
Specifications
Specifications
| Chemical Name or Material | LY3009120 |
| CAS | 1454682-72-4 |
| Color | Earth Yellow |
| Purity Grade Notes | Research |
| Molecular Formula | C23H29FN6O |
| Quantity | 100 mg |
| Synonym | DP-4978 |
| Solubility Information | DMSO : ≥ 38 mg/mL (89.51 mM) |
| SMILES | O=C(NC1=CC(C2=CC3=CN=C(NC)N=C3N=C2C)=C(C)C=C1F)NCCC(C)(C)C |
| Molecular Weight (g/mol) | 424.51 |
| Show More |
Research purposes only
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?