missing translation for 'onlineSavingsMsg'
Learn More
Learn More
LY2940094, MedChemExpress
Description
LY2940094 (BTRX-246040) is a potent, selective and orally available nociceptin receptor (NOP receptor) antagonist with high affinity (Ki=0.105 nM) and antagonist potency (Kb=0.166 nM). LY2940094 reduces ethanol self-administration in animal models.
Specifications
Specifications
| Chemical Name or Material | LY2940094 |
| CAS | 1307245-86-8 |
| Color | White |
| Purity Grade Notes | Research |
| Molecular Formula | C22H23ClF2N4O2S |
| Quantity | 25 mg |
| Synonym | BTRX-246040 |
| Solubility Information | DMSO : 41.67 mg/mL (86.64 mM; Need ultrasonic) |
| SMILES | OCC1=CC=CN=C1N2N=C(C)C(CN3CCC4(C5=C(C=C(Cl)S5)C(F)(F)CO4)CC3)=C2 |
| Molecular Weight (g/mol) | 480.96 |
| Show More |
Research purposes only
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?