missing translation for 'onlineSavingsMsg'
Learn More

STEMCELL Technologies LY294002

Product Code. 17140032
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 mg
25 mg
Unit Size:
25mg
5mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
17140032 5 mg 5mg
17150032 25 mg 25mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 17140032 Supplier STEMCELL Technologies Supplier No. 72152
 Restricted Item
Estimated Shipment: 10-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

PI3K/AKT pathway inhibitor; Inhibits PI3K

  • LY294002 is a PI3K inhibitor that has greater potency and selectivity than quercetin, the structure on which it is based
  • LY294002 inhibits PI3K (IC50 = 1.4 μM) and also shows activity against CK2, but not PI4K, EGFR, PDGFR, MAPK, PKA, or PKC
  • (Davies et al.), Vlahos et al.)
  • Suppresses proliferation and self-renewal of mouse embryonic stem (ES) cells (Lianguzova et al.), Paling et al.)
  • Promotes differentiation to insulin-producing cells from mouse ES cells (Hori et al.)
  • Inhibits myotube formation from myoblasts (Coolican et al.), Jiang et al.)

Specifications

CAS 154447-36-6
Quantity 5 mg
Molecular Formula C19H17NO3
Purity ≥98%
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.