missing translation for 'onlineSavingsMsg'
Learn More
Learn More
LY2811376, MedChemExpress
Description
LY2811376 is the first orally available non-peptidic β-secretase (BACE1) inhibitor with IC50 of 239 nM-249 nM, that acts to decrease Aβ secretion with EC50 of 300 nM, and demonstrates to have 10-fold selectivity towards BACE1 over BACE2, and more than 50-fold inhibition over other aspartic proteases including cathepsin D, pepsin, or renin.
Specifications
Specifications
| Chemical Name or Material | LY2811376 |
| CAS | 1194044-20-6 |
| Color | Earth Yellow |
| Purity Grade Notes | Research |
| Molecular Formula | C15H14F2N4S |
| Quantity | 100 mg |
| Solubility Information | DMSO : ≥ 31 mg/mL (96.77 mM) |
| SMILES | FC1=C(C2=CN=CN=C2)C=C([C@]3(C)CCSC(N3)=N)C(F)=C1 |
| Molecular Weight (g/mol) | 320.36 |
| Formula Weight | 320.36 |
| Show More |
Research purposes only
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?