missing translation for 'onlineSavingsMsg'
Learn More

LY2562175, MedChemExpress

Product Code. p-200098551
Encompass_Preferred
Click to view available options
Quantity:
5 mg
10 mg
50 mg
Unit Size:
10mg
50mg
5mg
This item is not returnable. View return policy

Product Code. 16490257

Brand: Medchem Express HY103704

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress LY2562175 is a potent and selective FXR agonist, with an EC50 of 193 nM.

LY2562175 is a potent and selective FXR agonist, with an EC50 of 193 nM.

Specifications

Chemical Name or Material LY2562175
CAS 1103500-20-4
Color White
Purity Grade Notes Research
Molecular Formula C28H27Cl2N3O4
Quantity 50 mg
Solubility Information DMSO : 62.5 mg/mL (115.65 mM; Need ultrasonic)
SMILES O=C(C1=CN(C)C2=C1C=CC(N3CCC(OCC4=C(C5CC5)ON=C4C6=C(Cl)C=CC=C6Cl)CC3)=C2)O
Molecular Weight (g/mol) 540.44
Formula Weight 540.44
Percent Purity 98.39%
Grade Research
Physical Form Solid
For Use With (Application) Neuroscience-Neuromodulation
Show More Show Less

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.