missing translation for 'onlineSavingsMsg'
Learn More
Learn More
LY2510924, MedChemExpress
Description
LY2510924 is a potent and selective CXCR4 antagonist that blocks SDF-1 binding to CXCR4 with an IC50 of 0.079 nM.
Specifications
Specifications
| Chemical Name or Material | LY2510924 |
| CAS | 1088715-84-7 |
| Color | White |
| Purity Grade Notes | Research |
| Molecular Formula | C62H88N14O10 |
| Quantity | 50 mg |
| Solubility Information | DMSO : ≥ 125 mg/mL (105.09 mM) |
| SMILES | O=C(NCC(N[C@H](CCC(N[C@H]1CC2=CC=CC=C2)=O)C(N[C@H](C(N)=O)CCCCNC(C)C)=O)=O)[C@@H](NC([C@@](NC([C@@H](NC([C@H](NC1=O)CC3=CC=C(O)C=C3)=O)CCCCNC(C)C)=O)([H])CCCNC(N)=N)=O)CC4=CC(C=CC=C5)=C5C=C4 |
| Molecular Weight (g/mol) | 1189.45 |
| Formula Weight | 1189.45 |
| Show More |
Research purposes only
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?