missing translation for 'onlineSavingsMsg'
Learn More

LY2452473, MedChemExpress

Product Code. 16491097
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 mg
10 mg
50 mg
100 mg
Unit Size:
100mg
10mg
50mg
5mg
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
16491097 50 mg 50mg
16417597 5 mg 5mg
16415636 100 mg 100mg
16404848 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Product Code. 16491097 Supplier Medchem Express Supplier No. HY114530
 Restricted Item
Estimated Shipment: 09-10-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

MedChemExpress LY2452473 is an orally bioavailable, selective androgen receptor modulator (SARM).

LY2452473 is an orally bioavailable, selective androgen receptor modulator (SARM).

Specifications

Chemical Name or Material LY2452473
CAS 1029692-15-6
Color Off-White
Purity Grade Notes Research
Molecular Formula C22H22N4O2
Quantity 50 mg
Solubility Information DMSO : 62.5 mg/mL (166.92 mM; Need ultrasonic)
SMILES O=C(N[C@@H]1CC(N(C2=C3C=C(C=C2)C#N)CC4=NC=CC=C4)=C3C1)OC(C)C
Molecular Weight (g/mol) 374.44
Formula Weight 374.44
Percent Purity 98.13%
Grade Research
Physical Form Solid
For Use With (Application) Cancer-programmed cell death
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.