missing translation for 'onlineSavingsMsg'
Learn More

LY223982, MedChemExpress

Product Code. 16487407
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 mg
Unit Size:
5mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
16487407 5 mg 5mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 16487407 Supplier Medchem Express Supplier No. HY112737
 Restricted Item
Estimated Shipment: 09-10-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

MedChemExpress LY223982 is a potent and specific inhibitor of leukotriene B4 receptor, with an IC50 of 13.2 nM against [3H]LTB4 binding to LTB4 receptor.

LY223982 is a potent and specific inhibitor of leukotriene B4 receptor, with an IC50 of 13.2 nM against [3H]LTB4 binding to LTB4 receptor.

Specifications

Chemical Name or Material LY223982
CAS 117423-74-2
Color White
Purity Grade Notes Research
Molecular Formula C30H30O7
Quantity 5 mg
Synonym CGS23131 SKF107324
Solubility Information DMSO : 100 mg/mL (198.98 mM; Need ultrasonic)
SMILES O=C(O)CCC1=CC(C(C2=CC=CC(C(O)=O)=C2)=O)=CC=C1OCCCC/C=C/C3=CC=C(OC)C=C3
Molecular Weight (g/mol) 502.56
Formula Weight 502.56
Percent Purity 97.02%
Grade Research
Physical Form Powder
For Use With (Application) COVID-19-immunoregulation
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.