missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Ly-6G Rat, Qfluor 630, Clone: RB6-8C5, enQuireBio™
Rat Monoclonal Antibody
£164.00 - £237.00
Specifications
Antigen | Ly-6G |
---|---|
Clone | RB6-8C5 |
Concentration | 0.2mg/mL |
Applications | Flow Cytometry |
Classification | Monoclonal |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15932468
|
EnquireBio
QAB77-QF630-100UG |
100 μg |
£164.00
100µg |
Estimated Shipment: 13-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15942468
|
EnquireBio
QAB77-QF630-500UG |
500 μg |
£237.00
500µg |
Estimated Shipment: 13-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Specifications
Ly-6G | |
0.2mg/mL | |
Monoclonal | |
Rat | |
10 mM NaH2PO4, 150 mM NaCl, 0.09% NaN3, 0.1% gelatin, pH7.2 | |
17067 | |
IgG2b κ | |
Protein A/G Purified |
RB6-8C5 | |
Flow Cytometry | |
Qfluor 630 | |
RUO | |
NP_001238984 | |
Ly6c1 | |
Primary | |
Store at 2-4°C. |