missing translation for 'onlineSavingsMsg'
Learn More

Ly-6G/Ly-6C Monoclonal Antibody (RB6-8C5), Alexa Fluor™ 488, eBioscience™, Invitrogen™

Rat Monoclonal Antibody

Brand:  Affymetrix eBioscience 53-5931-80

Code : Z2

Additional Details : Weight : 0.25000kg

 View more versions of this product

Product Code. 15322050

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products »


Description

Description

Description: The RB6-8C5 monoclonal antibody reacts with mouse Ly-6G, a 21-25 kDa protein also known as the myeloid differentiation antigen Gr-1. A GPI-linked protein, Gr-1 is expressed by the myeloid lineage in a developmentally regulated manner in the bone marrow. While monocytes only express Gr-1 transiently during their bone marrow development, the expression of Gr-1 on bone marrow granulocytes as well as on peripheral neutrophils is a good marker for these populations. eBioscience testing indicates that in the bone marrow and lysed whole blood, the antibody clone RB6-8C5 also stains cells that express the highest levels of Ly6c (as defined by staining with antibody clone HK1.4). It is recommended that 1A8-Ly6G (cat. 9668) be used when looking at Ly-6G specific targets. Applications Reported: This RB6-8C5 antibody has been reported for use in flow cytometric analysis. Applications Tested: This RB6-8C5 antibody has been tested by flow cytometric analysis of mouse bone marrow and splenocyte suspensions. This can be used at less than or equal to 0.125 μg per test. A test is defined as the amount (μg) of antibody that will stain a cell sample in a final volume of 100 μL. Cell number should be determined empirically but can range from 10^5 to 10^8 cells/test. It is recommended that the antibody be carefully titrated for optimal performance in the assay of interest. Excitation: 488 nm; Emission: 519 nm; Laser: Blue Laser.

Filtration: 0.2 μm post-manufacturing filtered. Granulocytes are a category of white blood cells characterized by the presence of granules in their cytoplasm. They are also called polymorphonuclear leukocytes (PMN or PML) because of the varying shapes of the nucleus, which is usually lobed into three segments. In common parlance, the term polymorphonuclear leukocyte often refers specifically to neutrophil granulocytes, the most abundant of the granulocytes. Granulocytes or PMN are released from the bone marrow by the regulatory complement proteins.
Specifications

Specifications

Ly-6G/Ly-6C
Monoclonal
0.5 mg/mL
PBS with 0.1% gelatin and 0.09% sodium azide; pH 7.2
P0CW03, P35461, P0CW02
Ly6c1, Ly6c2, Ly6g
Affinity chromatography
RUO
17067, 546644, 100041546
4° C, store in dark, DO NOT FREEZE!
Liquid
Flow Cytometry
RB6-8C5
Alexa Fluor 488
Ly6c1
Gr-1, Gr1, Ly-6G
Rat
25 μg
Primary
Mouse
Antibody
IgG2b κ
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

For Research Use Only.