missing translation for 'onlineSavingsMsg'
Learn More

Ly-6G/Ly-6C Antibody (RB6-8C5), DyLight 680, Novus Biologicals™

Product Code. 18338933
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.10mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18338933 0.1 mL 0.10mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18338933 Supplier Bio-Techne Supplier No. NBP311979FR

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rat Monoclonal Antibody

Ly-6G/Ly-6C Monoclonal antibody specifically detects Ly-6G/Ly-6C in Mouse samples. It is validated for Western Blot, Flow Cytometry, Immunohistochemistry
TRUSTED_SUSTAINABILITY

Specifications

Antigen Ly-6G/Ly-6C
Applications Western Blot, Flow Cytometry, Immunohistochemistry
Classification Monoclonal
Clone RB6-8C5
Conjugate DyLight 680
Dilution Western Blot, Flow Cytometry, Immunohistochemistry
Formulation 50mM Sodium Borate
Gene Alias GR, Gr-1, Ly-6, Ly6G
Host Species Rat
Immunogen This antibody was raised in Lou-M rats using a popliteal lymph node immunisation and fusion protocol
Purification Method Protein A purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Dendritic Cell Markers, Inflammation, Myeloid Cell Markers, Myeloid derived Suppressor Cell
Primary or Secondary Primary
Gene ID (Entrez) 546644
Target Species Mouse
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG2a κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.