missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Ly-6G/Ly-6C Antibody (RB6-8C5), Allophycocyanin, Novus Biologicals™
Rat Monoclonal Antibody
Brand: Novus Biologicals NBP2-00372-100UL
This item is not returnable.
View return policy
Description
Ly-6G/Ly-6C Monoclonal specifically detects Ly-6G/Ly-6C in Mouse samples. It is validated for Flow Cytometry.
Specifications
| Ly-6G/Ly-6C | |
| Monoclonal | |
| 0.20 mg/mL | |
| Flow Cytometry 1:10-1:1000 | |
| LY6G | |
| Normal mouse bone marrow. | |
| Primary | |
| Studies indicate that in the bone marrow and lysed whole blood, the antibody clone RB6-8C5 also reacts with an additional subpopulation of Ly-6C high cells. | |
| Store at 4°C. Do not freeze. |
| Flow Cytometry | |
| RB6-8C5 | |
| APC | |
| PBS with 0.05% sodium azide | |
| Rat | |
| 100 μL | |
| 546644 | |
| Mouse | |
| IgG2b κ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction