missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Ly-6E Recombinant Rabbit Monoclonal Antibody (9D8G1)

Rabbit Recombinant Monoclonal Antibody
Brand: Invitrogen™ MA557962
This item is not returnable.
View return policy
Description
Immunogen sequence: LMCFSCLNQK SNLYCLKPTI CSDQDNYCVT VSASAGIGNL VTFGHSLSKT CSPACPIPEG VNVGVASMGI SCCQSFLCNF.
Sca1, also known as Ly6A/E, is a member of the Ly6 multigene family of type V glycophosphatidylinositol anchored cell surface proteins. It is expressed on multipotent hematopoietic stem cells in bone marrow of mice with both the Ly6.1 and Ly6.2 allotypes. In the periphery, Sca1 exhibits a pattern of expression which is based on differences between the two allotypes. Ly6.1 strains (e.g., A, BALB/c, CBA, C3H/He, DBA/1, NZB) have few Sca1+ resting peripheral lymphocytes, whereas Ly6.2 strains (e.g., AKR, C57BL, C57BR, C57L, DBA/2, PL, SJL, SWR, 129) have relatively high numbers of Sca1+ lymphocytes. The expression of Sca1 is dramatically upregulated in all strains upon cellular activation.
Specifications
| Ly-6E | |
| Recombinant Monoclonal | |
| 1.34 mg/mL | |
| PBS with 0.05% BSA, 50% glycerol and 0.09% sodium azide; pH 7.3 | |
| Q16553 | |
| LY6E | |
| Recombinant protein (or fragment). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
| ELISA, Western Blot | |
| 9D8G1 | |
| Unconjugated | |
| LY6E | |
| 9804; locus A/E; Locus AE; Ly67; Ly6A; Ly6e; ly-6E; lymphocyte antigen 6 complex, locus E; lymphocyte antigen 6E; retinoic acid induced gene E; retinoic acid-induced gene E protein; RIGE; RIG-E; SCA2; SCA-2; Stem cell antigen 2; thymic shared antigen 1; Tsa1; Tsa-1 | |
| Rabbit | |
| Protein A | |
| RUO | |
| 4061 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction