missing translation for 'onlineSavingsMsg'
Learn More

Ly-6E Antibody, Novus Biologicals™

Product Code. 18398427 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mg
Unit Size:
0.01mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18398427 0.1 mg 0.01mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18398427 Supplier Novus Biologicals Supplier No. H00004061D01P

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

Ly-6E Polyclonal antibody specifically detects Ly-6E in Human samples. It is validated for Western Blot
TRUSTED_SUSTAINABILITY

Specifications

Antigen Ly-6E
Applications Western Blot
Classification Polyclonal
Conjugate Unconjugated
Formulation PBS (pH 7.4)
Gene Accession No. NP_002337.1
Gene Alias lymphocyte antigen 6 complex, locus E, Retinoic acid-induced gene E protein, RIGE9804, RIG-Ely-6E, SCA2Ly-6E, SCA-2lymphocyte antigen 6E, Stem cell antigen 2, Thymic shared antigen 1, TSA1, TSA-1retinoic acid induced gene E
Host Species Rabbit
Immunogen LY6E (NP_002337.1, 1 a.a. - 131 a.a.) full-length human protein. MKIFLPVLLAALLGVERASSLMCFSCLNQKSNLYCLKPTICSDQDNYCVTVSASAGIGNLVTFGHSLSKTCSPACPIPEGVNVGVASMGISCCQSFLCNFSAADGGLRASVTLLGAGLLLSLLPALLRFGP
Purification Method Protein A purified
Quantity 0.1 mg
Regulatory Status RUO
Research Discipline Signal Transduction
Primary or Secondary Primary
Gene ID (Entrez) 4061
Target Species Human
Content And Storage Aliquot and store at -20°C or -80°C. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.