missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Ly-6A Recombinant Rabbit Monoclonal Antibody (JM53-11)

Rabbit Recombinant Monoclonal Antibody
Brand: Invitrogen™ MA541224
This item is not returnable.
View return policy
Description
Ly-6A Recombinant Monoclonal Antibody for Western Blot, IHC (P)
LY6A is included in a family of proteins that are highly homologous, low molecular weight glycophosphotidylinositol-linked proteins expressed on hematopoietic cells and lymphoid cells.
Specifications
| Ly-6A | |
| Recombinant Monoclonal | |
| 1 mg/mL | |
| TBS with 0.05% BSA, 40% glycerol and 0.05% sodium azide; pH 7.4 | |
| P05533 | |
| Ly6a | |
| recombinant protein. | |
| 100 μL | |
| Primary | |
| Mouse | |
| Antibody | |
| IgG |
| Immunohistochemistry (Paraffin), Western Blot | |
| JM53-11 | |
| Unconjugated | |
| Ly6a | |
| Ly6; Ly6a; Ly-6A.2; Ly-6A.2/Ly-6E.1; Ly-6A/E; Ly-6E.1; lymphocyte antigen 6 complex, locus A; lymphocyte antigen 6A-2/6E-1; Sca1; SCA-1; Stem cell antigen 1; TAP; T-cell-activating protein | |
| Rabbit | |
| Protein A | |
| RUO | |
| 110454 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
Corrección del contenido de un producto
Proporcione sus comentarios sobre el contenido del producto rellenando el siguiente formulario.
Título del producto
¿Detecta una oportunidad de mejora?Comparta una corrección de contenido