Learn More
Ly-6A/E (Sca-1) Monoclonal Antibody (D7), eBioscience™, Invitrogen™

Rat Monoclonal Antibody
£38.81 - £195.00
Specifications
| Antigen | Ly-6A/E (Sca-1) |
|---|---|
| Clone | D7 |
| Concentration | 0.5 mg/mL |
| Classification | Monoclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
15237657
|
Invitrogen
14-5981-81 |
50 μg | |||||||
15247657
![]() |
Invitrogen™
14-5981-82 |
100 μg |
£78.50
100µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
15257657
|
Invitrogen™
14-5981-85 |
500 μg |
£195.00
500µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Description: The D7 monoclonal antibody reacts with mouse Sca-1, an 18 kDa member of the Ly-6 family of GPI-linked surface proteins. D7 reacts with both Ly-6E.1 and Ly-6A.2 molecules expressed by mouse hematopoietic stem cells, myeloid population and peripheral T and B cells. Non-activated splenocytes from Ly-6.2 strains including C57BL, SJL, 129, AKR, and others demonstrate higher frequency of D7-positive cells compared to the Ly-6.1 strains such as BALB/c, C3H, NZB and others. Sca-1 is expressed at high levels upon activation regardless of the Ly-6 haplotype. The Ly-6 family is involved in regulation and function of T cell activation. Sca-1 is a major phenotypic marker for mouse hematopoietic progenitor/stem cell subset. Applications Reported: This D7 antibody has been reported for use in flow cytometric analysis, immunoprecipitation, and western blotting. Applications Tested: This D7 antibody has been tested by flow cytometric analysis of mouse bone marrow cells. This can be used at less than or equal to 0.5 μg per test. A test is defined as the amount (μg) of antibody that will stain a cell sample in a final volume of 100 μL. Cell number should be determined empirically but can range from 10^5 to 10^8 cells/test. It is recommended that the antibody be carefully titrated for optimal performance in the assay of interest. (Fluorochrome-conjugated D7 is recommended for use in flow cytometry.).
Stem Cell Antigen-1 (Sca-1) is a member of the lymphocyte antigen 6 (Ly-6) family. It is expressed on multipotent hematopoietic stem cells (HSC) and is used as a marker of HSC in mice. Sca-1 is used in combination with lineage depletion antibodies to identify murine HSC. Sca-1 can be found in fetal liver, adult bone marrow, and adult peripheral blood and spleen. Sca-1 may be involved in the regulation of B and T cell activation.Specifications
| Ly-6A/E (Sca-1) | |
| 0.5 mg/mL | |
| Unconjugated | |
| Rat | |
| Mouse | |
| 110454, 17069 | |
| IgG2a κ | |
| Affinity chromatography | |
| Antibody |
| D7 | |
| Monoclonal | |
| Liquid | |
| RUO | |
| P05533, Q64253 | |
| Ly6a, Ly6e | |
| Primary | |
| 4° C |
For Research Use Only.
Your input is important to us. Please complete this form to provide feedback related to the content on this product.