missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Ly-6A/E (Sca-1) Monoclonal Antibody (D7), Brilliant Violet™ 605, eBioscience™, Invitrogen™

Rat Monoclonal Antibody
£167.00 - £341.00
Specifications
| Antigen | Ly-6A/E (Sca-1) |
|---|---|
| Clone | D7 |
| Concentration | 0.2 mg/mL |
| Applications | Flow Cytometry |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
17846226
|
Invitrogen™
406-5981-80 |
25 μg |
£167.00
25µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
17826216
![]() |
Invitrogen™
406-5981-82 |
100 μg |
£341.00
100µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Stem Cell Antigen-1 (Sca-1) is a member of the lymphocyte antigen 6 (Ly-6) family. It is expressed on multipotent hematopoietic stem cells (HSC) and is used as a marker of HSC in mice. Sca-1 is used in combination with lineage depletion antibodies to identify murine HSC. Sca-1 can be found in fetal liver, adult bone marrow, and adult peripheral blood and spleen. Sca-1 may be involved in the regulation of B and T cell activation.Specifications
| Ly-6A/E (Sca-1) | |
| 0.2 mg/mL | |
| Monoclonal | |
| Liquid | |
| RUO | |
| PBS with BSA and 0.09% sodium azide; pH 7.2 | |
| 9804; locus A/E; Locus AE; Ly6; Ly67; Ly6a; Ly-6A.2; Ly-6A.2/Ly-6E.1; Ly6A/E; Ly-6A/E; ly6a-e; Ly6e; Ly-6E; Ly-6E.1; lymphocyte antigen 6 complex; lymphocyte antigen 6 complex, locus A; lymphocyte antigen 6 complex, locus E; lymphocyte antigen 6A-2/6E-1; Lymphocyte antigen 6E; RIG-E; Sca1; Sca-1; Sca-2; stem cell antigen 1; Stem cell antigen 2; TAP; T-cell-activating protein; thymic shared antigen 1; Tsa1; TSA-1 | |
| Ly6a, Ly6e | |
| Primary | |
| 4°C, store in dark, DO NOT FREEZE! |
| D7 | |
| Flow Cytometry | |
| Brilliant Violet 605 | |
| Rat | |
| Mouse | |
| P05533, Q64253 | |
| 110454, 17069 | |
| IgG2a κ | |
| Affinity chromatography |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title