Learn More
Ly-6A/E Rat anti-Mouse, PE, Clone: D7, BD
Rat Monoclonal Antibody
Brand: BD Biosciences 553108
Description
The D7 antibody reacts with Ly-6A.2 and Ly-6E.1, which are allelic members of the Ly-6 multigene family. Sca-1 (Ly6A/E), a phosphatidylinositol-anchored protein of about 18kDa, is expressed on the multipotent hematopoietic stem cells (HSC) in the bone mar
Host Species: Rat
Clone: D7
Isotype: IgG2a κ
Species Reactivity: Mouse
Immunogen: IL-2-dependent mouse T-cell line CTL-L
Flow Cytometry
Specifications
| Ly-6A/E | |
| Monoclonal | |
| 0.2mg/mL | |
| Ly-6A/E; Lymphocyte antigen 6A-2/6E-1; Ly-6A.2/Ly-6E.1; Sca-1; TAP | |
| Rat | |
| Affinity Purified | |
| RUO | |
| Primary | |
| Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze. |
| Flow Cytometry | |
| D7 | |
| PE | |
| Aqueous buffered solution containing ≤0.09% sodium azide. | |
| IL-2-dependent mouse T-cell line CTL-L | |
| 0.2 mg | |
| Immunology | |
| Mouse | |
| IgG2a κ |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
For Research Use Only.