Learn More
Ly-6A/E Rat anti-Mouse, FITC, Clone: E13-161.7, BD
Rat Monoclonal Antibody
£66.54 - £319.93
Specifications
| Antigen | Ly-6A/E |
|---|---|
| Clone | E13-161.7 |
| Concentration | 0.5mg/mL |
| Applications | Flow Cytometry |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
15840749
|
BD Biosciences
561077 |
25 μg |
£68.50 £66.54 / Each Save £1.96 2% Off |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
15847158
|
BD Biosciences
553335 |
0.5 mg |
£324.00 £319.93 / Each Save £4.07 1% Off |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
The E13-161.7 antibody reacts with Ly-6A.2 and Ly-6E.1, which are allelic members of the Ly-6 multigene family. Sca1 (Ly-6A/E), a phosphatidylinositol-anchored protein of about 18 kDa, is expressed on the multipotent hematopoietic stem cell (HSC) in mice with both Ly-6 haplotypes. Sca-1+ HSC are found in the adult bone marrow and fetal liver, but not in the early embryo yolk sac or intraembryonic hematopoietic sites, and can be mobilized to the peripheral blood and spleen in the adult. In mice expressing the Ly-6.2 haplotype (e.g., AKR, C57BL, C57BR, C57L, C58, DBA/2, PL, SJL, SWR, 129), Ly-6A/E is also expressed on distinct subpopulations of bone marrow and peripheral B lymphocytes, myeloid cells, and thymic and peripheral T lymphocytes, on the earliest intrathymic T-cell precursor population, and in several non-hematopoietic tissues. Strains with the Ly-6.1 haplotype (e.g., A, BALB/c, CBA, C3H/He, DBA/1, NZB) have few Ly-6A/E+ resting peripheral lymphocytes, whereas activation of T cells from mice of both Ly-6 haplotypes leads to strong expression of the Sca-1 antigen. Studies with the D7 antibody (Cat. No. 557403) have demonstrated that Ly-6A/E may be involved in the regulation of B and T lymphocyte responses, and it appears to be required for T-cell receptor-mediated T-cell activation. Purified E13-161.7 mAb can block binding of FITC-conjugated D7 antibody (anti-Ly-6A/E, Cat. No. 557405) to mouse splenocytes, but purified mAb D7 (Cat. No. 557403) is unable to block binding of FITC-conjugated E13-161.7 antibody (Cat. No. 553335). Anti-Ly-6A/E (Sca-1) mAb may be used in combination with the Mouse Lineage Panel (Cat. No. 559971) to identify HSC.
This antibody is routinely tested by flow cytometric analysis. Other applications were tested at BD Biosciences Pharmingen during antibody development only or reported in the literature.
Specifications
| Ly-6A/E | |
| 0.5mg/mL | |
| Monoclonal | |
| Rat | |
| Mouse | |
| IgG2a κ | |
| Affinity Purified | |
| Sca-1 |
| E13-161.7 | |
| Flow Cytometry | |
| FITC | |
| RUO | |
| Aqueous buffered solution containing ≤0.09% sodium azide. | |
| Primary | |
| Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze. |
For Research Use Only.
Your input is important to us. Please complete this form to provide feedback related to the content on this product.