missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Ly-6A/E Rabbit anti-Mouse, Clone: 4N5K2, Novus Biologicals™
Rabbit Monoclonal Antibody
£150.00 - £386.00
Specifications
| Antigen | Ly-6A/E |
|---|---|
| Clone | 4N5K2 |
| Applications | Western Blot |
| Classification | Monoclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18351656
|
Novus Biologicals
NBP3-15949-20UL |
20 μg |
£150.00
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18377494
|
Novus Biologicals
NBP3-15949-100UL |
100 μg |
£386.00
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Ly-6A/E Monoclonal antibody specifically detects Ly-6A/E in Mouse samples. It is validated for Western BlotSpecifications
| Ly-6A/E | |
| Western Blot | |
| Unconjugated | |
| Rabbit | |
| Hematopoietic Stem Cell Markers, Stem Cell Markers | |
| PBS, 0.05% BSA, 50% glycerol, pH7.3 | |
| 110454 | |
| IgG | |
| Affinity purified |
| 4N5K2 | |
| Monoclonal | |
| Purified | |
| RUO | |
| Mouse | |
| Ly 6A/E, Ly6A.2, Ly6A/E, Ly6E.1, Lymphocyte antigen 6 complex locus A, Lymphocyte antigen 6 complex locus E, Retinoic acid induced gene E, RIGE, SCA1, SCA-1, SCA2, Stem cell antigen 1, Stem cell antigen 2, T cell activating protein, TAP, Thymic shared antigen 1, TSA1 | |
| A synthetic peptide corresponding to a sequence within amino acids 1-100 of mouse Ly-6A/E (P05533). | |
| Primary | |
| Store at -20°C. Avoid freeze-thaw cycles. |
Haben Sie Verbesserungsvorschläge?Übermitteln Sie eine inhaltliche Korrektur
Berichtigung von Produktinhalten
Bitte geben Sie uns Ihr Feedback zu den Produktinhalten, indem Sie das folgende Formular ausfüllen.
Name des Produkts