missing translation for 'onlineSavingsMsg'
Learn More

Ly-51 Rat anti-Mouse, FITC, Clone: 6C3, BD

Rat Monoclonal Antibody

Brand:  BD Biosciences 553160

 View more versions of this product

Product Code. 15817048

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products »

This item is not returnable. View return policy

Description

Description

The 6C3 antibody reacts with an epitope of the 6C3/BP-1 (Ly-51) glycoprotein cell-surface differentiation antigen, which was originally identified on pre-B cell lymphomas (spontaneous and chemical- or retrovirus- transformed, in vitro and in vivo). 6C3/BP

Host Species: Rat
Clone: 6C3
Isotype: IgG2a κ
Species Reactivity: Mouse
Immunogen: C57L mouse Pre-B lymphhoma cell line L1-2

Flow Cytometry

TRUSTED_SUSTAINABILITY
Specifications

Specifications

Ly-51
Monoclonal
0.5mg/mL
6C3/BP-1 Antigen
Rat
Affinity Purified
RUO
Primary
Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze.
Flow Cytometry
6C3
FITC
Aqueous buffered solution containing ≤0.09% sodium azide.
C57L mouse Pre-B lymphhoma cell line L1-2
0.5 mg
Immunology
Mouse
IgG2a κ
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.

For Research Use Only.