missing translation for 'onlineSavingsMsg'
Learn More

Ly-51 Rat anti-Mouse, FITC, Clone: 6C3, BD

Rat Monoclonal Antibody

Brand:  BD Biosciences 562057

 View more versions of this product

Product Code. 15801609

  • ONLINE EXCLUSIVE PRICE
    £158.00
    £78.15 / Each
    Save £79.85
    50% Off

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

The 6C3 antibody reacts with an epitope of the 6C3/BP-1 (Ly-51) glycoprotein cell-surface differentiation antigen, which was originally identified on pre-B cell lymphomas (spontaneous and chemical- or retrovirus- transformed, in vitro and in vivo). 6C3/BP-1 is a homodimer cell-surface glycoprotein with 140-kDa subunits which has been identified to possess aminopeptidase A (APA) activity. The same antigen is expressed at high levels on bone marrow stromal cell lines which support in vitro B lymphopoieses, on thymic dendritic cells and cortical epithelial cells, and on a wide variety of mouse and rat tissues known to possess APA activity. Subsets of normal bone marrow pre-B and B lymphocytes express low levels of Ly-51, which is rapidly up-regulated on the pre-B cells in the presence of IL-7. A role for the 6C3/BP-1 molecule in the IL-7-driven proliferation of B cell precursors has been postulated. However, B-cell abnormalities were not detected in Ly-51-deficient mice. Mature B lymphocytes, thymocytes, peripheral T lymphocytes, erythroid cells, and myeloid cells (with the exception of thymic dendritic cells) do not express Ly-51. The 6C3 antibody can be used to identify cortical epithelium in frozen sections of thymuses from normal, SCID, and TCR-transgenic mice. It is possible that the low level of 6C3/BP-1 antigen detected, by flow cytometry, on some thymocytes may be passively adsorbed from adjacent epithelial cells during preparation of the cell suspensions.

TRUSTED_SUSTAINABILITY
Specifications

Specifications

Ly-51
Monoclonal
0.5mg/mL
6C3/BP-1 Antigen
Rat
Affinity Purified
RUO
Mouse
IgG2a κ
Flow Cytometry
6C3
FITC
Aqueous buffered solution containing ≤0.09% sodium azide.
C57L mouse Pre-B lymphhoma cell line L1-2
50 μg
Primary
Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze.
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.

For Research Use Only.