Learn More
Ly-49G2 Monoclonal Antibody (eBio4D11 (4D11)), PerCP-eFluor™ 710, eBioscience™, Invitrogen™
Description
Description: The 4D11 monoclonal antibody reacts with Ly-49G2 (also known as LGL-1). The Ly-49 family of NK-cell receptors, members of the C-type lectin superfamily, are disulfide-linked type-II transmembrane protein homodimers with extracellular carbohydrate recognition domains, which bind to MHC class I alloantigens. The Ly-49 family members are expressed independently, such that an individual NK or T cell may display more than one class of Ly-49 receptor. Ly-49G2 is an inhibitory receptor expressed on subsets of natural killer (NK) cells, NK-T cells (in all strains tested), and on a population of memory CD8+ T cells (in C57BL/6 mice). The 4D11 antibody has been reported to cross react with the Ly-49AB6, Ly-49ABALB, and Ly-49T129/J inhibitory receptors and Ly-49LCBA/J activatingreceptors. The proportion of NK-T cells expressing Ly-49A and Ly-49G2 is higher in the thymus than in liver, and there is evidence that down-regulation of Ly-49 receptor expression is necessary for normal NK-T-cell development to occur. Applications Reported: This eBio4D11 (4D11) antibody has been reported for use in flow cytometric analysis. Applications Tested: This eBio4D11 (4D11) antibody has been tested by flow cytometric analysis of mouse spleen cells. This can be used at less than or equal to 0.125 μg per test. A test is defined as the amount (μg) of antibody that will stain a cell sample in a final volume of 100 μL.
Specifications
Specifications
| Antigen | Ly-49G2 |
| Applications | Flow Cytometry |
| Classification | Monoclonal |
| Clone | eBio4D11 (4D11) |
| Concentration | 0.2 mg/mL |
| Conjugate | PerCP-eFluor 710 |
| Formulation | PBS with 0.1% gelatin and 0.09% sodium azide; pH 7.2 |
| Gene | Klra7 |
| Gene Accession No. | Q60654 |
| Gene Alias | Killer cell lectin-like receptor 7, Lymphocyte antigen 49g, Ly-49g, T-cell surface glycoprotein Ly-49G, Klra7, Ly-49g, Ly49-g, Ly49g, Ly49g4 |
| Show More |
For Research Use Only.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.