Learn More
Ly-49C and Ly-49I Mouse anti-Mouse, FITC, Clone: 5E6, BD
Mouse Monoclonal Antibody
Brand: BD Biosciences 553276
Description
The 5E6 (also known as clone SW5E6) antibody reacts with Ly-49C[BALB], Ly-49C[B6], Ly-49C[NZB], and Ly-49I[B6], inhibitory receptors which are expressed on subsets of natural killer (NK) cells and NK-1.1+ (or DX5+) T lymphocytes (NK-T cells) in
Host Species: Mouse
Clone: 5E6
Isotype: IgG2a κ
Species Reactivity: Mouse
Immunogen: Activated mouse NK cells
Flow Cytometry
Specifications
| Ly-49C and Ly-49I | |
| Monoclonal | |
| 0.5mg/mL | |
| Aqueous buffered solution containing ≤0.09% sodium azide. | |
| Activated mouse NK cells | |
| 0.5 mg | |
| Immunology | |
| Mouse | |
| IgG2a κ |
| Flow Cytometry | |
| 5.00E+06 | |
| FITC | |
| Mouse | |
| Affinity Purified | |
| RUO | |
| Primary | |
| Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze. |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
For Research Use Only.