missing translation for 'onlineSavingsMsg'
Learn More

LY-411575, MedChemExpress

Product Code. 16488098
Encompass_Preferred
Click to view available options
Quantity:
5 mg
10 mg
50 mg
100 mg
Unit Size:
100mg
10mg
50mg
5mg
This item is not returnable. View return policy

Product Code. 16488098

Brand: Medchem Express HY50752

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress LY-411575 is a potent γ-secretase inhibitor with IC50 of 0.078 nM/0.082 nM (membrane/cell-based), and also inhibits Notch S3 cleavage with IC50 of 0.39 nM.

LY-411575 is a potent γ-secretase inhibitor with IC50 of 0.078 nM/0.082 nM (membrane/cell-based), and also inhibits Notch S3 cleavage with IC50 of 0.39 nM.

Specifications

Chemical Name or Material LY-411575
CAS 209984-57-6
Color White
Purity Grade Notes Research
Molecular Formula C26H23F2N3O4
Quantity 10 mg
Solubility Information DMSO : 33.33 mg/mL (69.51 mM; Need ultrasonic)
SMILES O=C1[C@@H](NC([C@@H](NC([C@H](C2=CC(F)=CC(F)=C2)O)=O)C)=O)C3=CC=CC=C3C(C=CC=C4)=C4N1C
Molecular Weight (g/mol) 479.48
Formula Weight 479.48
Percent Purity 98.0%
Grade Research
Physical Form Solid
For Use With (Application) Neuroscience-Neurodegeneration
Show More Show Less

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.