missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Tocris Bioscience™ LY 404187

Click to view available options
Quantity:
10 mg
50 mg
Unit Size:
10mg
50mg
Description
LY 404187 is a selective AMPA receptor positive allosteric modulator (IC50 values are 0.15, 0.21, 1.66 and 5.65 μM for GluA2i, GluA4i, GluA3i and GluA1i respectively). Potentiates glutamate and AMPA-evoked currents in pre frontal cortex (PFC) pyramidal neurons in vitro. Has no effect on NMDA- or kainate-evoked currents; also has no effect on K+ or Na+ ion channels. Potentiates PFC glutamatergic synaptic transmission in vitro and in vivo. Induces neurite growth in combination with (S)-AMPA (Cat. No. 0254) in SH-SY5Y human neuroblastoma cells in vitro. Also reduces MPTP-induced toxicity in mice. Brain penetrant.
Specifications
Specifications
| Chemical Name or Material | N-[2-(4′-Cyano[1,1′-biphenyl]-4-yl)propyl]-2-propanesulfonamide |
| CAS | 211311-95-4 |
| Quantity | 10 mg |
| Target | AMPA Receptor Modulators |
| Molecular Formula | C19H22N2O2S |
| Formula Weight | 342.46 |
| Purity | >98% |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction