missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Tocris Bioscience™ LY 367385

Click to view available options
Quantity:
10 mg
50 mg
Unit Size:
10mg
50mg
Description
A selective mGlu1a receptor antagonist, with an IC50 value of 8.8 μM for blockade of quisqualate-induced phosphoinositide hydrolysis vs. > 100 μM for mGlu5a, and negligible action on group II and III receptors.
Specifications
Specifications
| Quantity | 50 mg |
| Formula Weight | 209.2 |
| Purity | >99% |
| Molecular Weight (g/mol) | 209.201 |
| Chemical Name or Material | LY 367385 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction