missing translation for 'onlineSavingsMsg'
Learn More

LY 345899, MedChemExpress

Product Code. p-200097236
Encompass_Preferred
Click to view available options
Quantity:
5 mg
10 mg
25 mg
50 mg
100 mg
Unit Size:
100mg
10mg
25mg
50mg
5mg
This item is not returnable. View return policy

Product Code. 16483648

Brand: Medchem Express HY101943

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress LY 345899 is a Folate analog and is a methylene tetrahydrofolate dehydrogenase (MTHFD1; DC301) and MTHFD2 inbhibitor with IC50 values of 96 nM and 663 nM, respectively and a Ki of 18 nM for MTHFD1.

LY 345899 is a Folate analog and is a methylene tetrahydrofolate dehydrogenase (MTHFD1

Specifications

Chemical Name or Material LY 345899
Physical Form Solid
CAS 10538-99-5
Color Light Yellow
Quantity 10 mg
Purity Grade Notes Research
Molecular Formula C20H21N7O7
For Use With (Application) Cancer-programmed cell death
Solubility Information DMSO : 125 mg/mL (265.16 mM; Need ultrasonic)
SMILES O=C(O)[C@@H](NC(C1=CC=C(N2C(N3C4=C(NC(N)=NC4=O)NCC3C2)=O)C=C1)=O)CCC(O)=O
Molecular Weight (g/mol) 471.42
Formula Weight 471.42
Percent Purity 98.0%
Grade Research
Show More Show Less

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.