missing translation for 'onlineSavingsMsg'
Learn More

LY 345899, MedChemExpress

Product Code. 16444736
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 mg
10 mg
25 mg
50 mg
100 mg
Unit Size:
100mg
10mg
25mg
50mg
5mg
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
16444736 100 mg 100mg
16483648 10 mg 10mg
16446387 5 mg 5mg
16420047 50 mg 50mg
16402816 25 mg 25mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
This item is not returnable. View return policy
Product Code. 16444736 Supplier Medchem Express Supplier No. HY101943
 Restricted Item
Estimated Shipment: 18-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

MedChemExpress LY 345899 is a Folate analog and is a methylene tetrahydrofolate dehydrogenase (MTHFD1; DC301) and MTHFD2 inbhibitor with IC50 values of 96 nM and 663 nM, respectively and a Ki of 18 nM for MTHFD1.

LY 345899 is a Folate analog and is a methylene tetrahydrofolate dehydrogenase (MTHFD1

Specifications

Chemical Name or Material LY 345899
Physical Form Solid
CAS 10538-99-5
Color Light Yellow
Quantity 100 mg
Purity Grade Notes Research
Molecular Formula C20H21N7O7
For Use With (Application) Cancer-programmed cell death
Solubility Information DMSO : 125 mg/mL (265.16 mM; Need ultrasonic)
SMILES O=C(O)[C@@H](NC(C1=CC=C(N2C(N3C4=C(NC(N)=NC4=O)NCC3C2)=O)C=C1)=O)CCC(O)=O
Molecular Weight (g/mol) 471.42
Formula Weight 471.42
Percent Purity 98.0%
Grade Research
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.