missing translation for 'onlineSavingsMsg'
Learn More

LY 344864 S-enantiomer, MedChemExpress

Product Code. 16430206
Encompass_Preferred
Change view
Click to view available options
Quantity:
2 mg
5 mg
Unit Size:
2mg
5mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
16430206 2 mg 2mg
16428517 5 mg 5mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 16430206 Supplier Medchem Express Supplier No. HY13788A
 Restricted Item
Estimated Shipment: 16-10-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

MedChemExpress LY 344864 S-enantiomer is the S-enantiomer of LY344864. LY344864 is a 5-HT1F receptor agonist.

LY 344864 S-enantiomer is the S-enantiomer of LY344864. LY344864 is a 5-HT1F receptor agonist.

Specifications

Chemical Name or Material LY 344864 S-enantiomer
CAS 186544-27-4
Color Light Yellow
Purity Grade Notes Research
Molecular Formula C21H22FN3O
Quantity 2 mg
SMILES O=C(NC1=CC2=C(NC3=C2C[C@@H](N(C)C)CC3)C=C1)C4=CC=C(F)C=C4
Molecular Weight (g/mol) 351.42
Formula Weight 351.42
Percent Purity 98.0%
Grade Research
Physical Form Solid
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.