missing translation for 'onlineSavingsMsg'
Learn More

Tocris Bioscience™ LY 341495 disodium salt
GREENER_CHOICE

Product Code. 18055652
Click to view available options
Quantity:
1 mg
10 mg
Unit Size:
10mg
1mg
This item is not returnable. View return policy

Product Code. 18055652

Brand: Tocris Bioscience™ 4062/10

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Potent and selective group II mGlu antagonist; disodium salt of LY 341495 (Cat. No. 1209)

Highly potent and selective group II metabotropic glutamate receptor antagonist. Disodium salt of LY 341495 (Cat. No. 1209).
TRUSTED_SUSTAINABILITY

Chemical Identifiers

Molecular Formula C20H17NNa2O5

Specifications

Quantity 10 mg
Formula Weight 397.33
Purity >97%
Chemical Name or Material LY 341495 disodium salt
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.