missing translation for 'onlineSavingsMsg'
Learn More

LY 303511 hydrochloride, MedChemExpress

Product Code. 16412797
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 mg
10 mg
50 mg
Unit Size:
10mg
50mg
5mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16412797 50 mg 50mg
16496708 10 mg 10mg
16459267 5 mg 5mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 16412797 Supplier Medchem Express Supplier No. HY15643A

Please to purchase this item. Need a web account? Register with us today!

This item requires a price update prior to being ordered.

Request a Price for this item here.

This item is not returnable. View return policy

MedChemExpress LY 303511 hydrochloride is a structural analogue of LY294002. LY303511 does not inhibit PI3K. LY303511 enhances TRAIL sensitivity of SHEP-1 neuroblastoma cells. LY303511 reversibly blocks K+ currents (IC50=64.6±9.1 μM) in MIN6 insulinoma cells.

LY 303511 hydrochloride is a structural analogue of LY294002. LY303511 does not inhibit PI3K. LY303511 enhances TRAIL sensitivity of SHEP-1 neuroblastoma cells. LY303511 reversibly blocks K+ currents (IC50=64.6±9.1 μM) in MIN6 insulinoma cells.

Specifications

Chemical Name or Material LY 303511 hydrochloride
CAS 2070014-90-1
Color Off-White
Purity Grade Notes Research
Molecular Formula C19H19ClN2O2
Quantity 50 mg
Solubility Information DMSO : 25 mg/mL (72.92 mM; Need ultrasonic) ∣H2O : 1 mg/mL (2.92 mM; Need ultrasonic)
SMILES O=C1C=C(N2CCNCC2)OC3=C(C4=CC=CC=C4)C=CC=C13.Cl
Molecular Weight (g/mol) 342.82
Formula Weight 342.82
Percent Purity 98.78%
Grade Research
Physical Form Solid
For Use With (Application) Cancer-Kinase/protease
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.