missing translation for 'onlineSavingsMsg'
Learn More
Learn More
LY 303511 hydrochloride, MedChemExpress
Click to view available options
Quantity:
5 mg
10 mg
50 mg
Unit Size:
10mg
50mg
5mg
Description
LY 303511 hydrochloride is a structural analogue of LY294002. LY303511 does not inhibit PI3K. LY303511 enhances TRAIL sensitivity of SHEP-1 neuroblastoma cells. LY303511 reversibly blocks K+ currents (IC50=64.6±9.1 μM) in MIN6 insulinoma cells.
Specifications
Specifications
| Chemical Name or Material | LY 303511 hydrochloride |
| CAS | 2070014-90-1 |
| Color | Off-White |
| Purity Grade Notes | Research |
| Molecular Formula | C19H19ClN2O2 |
| Quantity | 10 mg |
| Solubility Information | DMSO : 25 mg/mL (72.92 mM; Need ultrasonic) ∣H2O : 1 mg/mL (2.92 mM; Need ultrasonic) |
| SMILES | O=C1C=C(N2CCNCC2)OC3=C(C4=CC=CC=C4)C=CC=C13.Cl |
| Molecular Weight (g/mol) | 342.82 |
| Formula Weight | 342.82 |
| Show More |
Research purposes only
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction