missing translation for 'onlineSavingsMsg'
Learn More

LY 3000328, MedChemExpress

Product Code. 16452697
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 mg
10 mg
50 mg
Unit Size:
10mg
50mg
5mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16452697 50 mg 50mg
16436608 10 mg 10mg
16419177 5 mg 5mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 16452697 Supplier Medchem Express Supplier No. HY15533

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress LY 3000328 is a potent and selective Cathepsin S (Cat S) inhibitor with IC50s of 7.7 and 1.67 nM for hCat S and mCat S, respectively.

LY 3000328 is a potent and selective Cathepsin S (Cat S) inhibitor with IC50s of 7.7 and 1.67 nM for hCat S and mCat S, respectively.

Specifications

Chemical Name or Material LY 3000328
CAS 1373215-15-6
Color White
Purity Grade Notes Research
Molecular Formula C25H29FN4O5
Quantity 50 mg
Solubility Information DMSO : ≥ 50 mg/mL (103.19 mM)
SMILES O=C(C1=CC=C(F)C=C1)N[C@@H]2[C@@H](OC(NC)=O)COC3=CC=C(N4CCN(C5COC5)CC4)C=C32
Molecular Weight (g/mol) 484.52
Formula Weight 484.52
Percent Purity 98.12%
Grade Research
Physical Form Solid
For Use With (Application) COVID-19-immunoregulation
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.