missing translation for 'onlineSavingsMsg'
Learn More

Tocris Bioscience™ LY 272015 hydrochloride
GREENER_CHOICE

Product Code. 30001446
Change view
Click to view available options
Quantity:
10 mg
50 mg
Unit Size:
10mg
50mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30001446 10 mg 10mg
18084322 50 mg 50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30001446 Supplier Tocris Bioscience™ Supplier No. 3077/10
 Restricted Item
Estimated Shipment: 03-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High affinity 5-HT2B antagonist; orally active

High affinity 5-HT2B receptor antagonist that displays selectivity over 5-HT2A and 5-HT2C receptors (Ki values are 0.75, 21.63 and 28.7 nM for 5-HT2B, 5-HT2C and 5-HT2A receptors respectively). Orally active.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 172895-15-7
Molecular Formula C21H25ClN2O2

Specifications

Quantity 10 mg
Formula Weight 372.89
Purity >99%
Molecular Weight (g/mol) 372.893
Chemical Name or Material LY 272015 hydrochloride
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.